
1-Nitro-4-(trifluoromethoxy)benzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:713-65-5
:
Formula:C₇H₄F₃NO₃
MDL Number:MFCD00040302
:207.11 g/mol
Boiling Pt:87…89 °C/15mmHg
Melting Pt:15 °C
Density:1.447 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together