
1-(Chloromethyl)-2-(trifluoromethoxy)benzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:116827-40-8
:
Formula:C₈H₆ClF₃O
MDL Number:MFCD00221456
:210.58 g/mol
Boiling Pt:68…70 °C/15mmHg
:GHS05
Storage Temperature:Ambient
Frequently Bought Together