Order Entry
Czech Republic
ContactUsLinkComponent
 
Corrosive
Irritant
4-(Trifluoromethoxy)benzylamine 96%
Catalog # APOSPC7438S-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
4-(Trifluoromethoxy)benzylamine 96%
Catalog # APOSPC7438S-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :191.15 g/mol
Boiling Pt:57…59 °C/10mmHg
Density:1.252 g/cm³ (25 °C)
Flash Pt:77 °C
Storage Temperature:Ambient
EINECS:300-040-1
UN:2735
ADR:8,III
 :GHS05
 :GHS07
CAS Number:93919-56-3
Formula:C₈H₈F₃NO
MDL Number:MFCD00061237