
2-(Trifluoromethoxy)benzylamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175205-64-8
:
Formula:C₈H₈F₃NO
MDL Number:MFCD00085176
:191.15 g/mol
Boiling Pt:77 °C/12mmHg
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together