
2-(Trifluoromethoxy)benzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:162046-61-9
:
Formula:C₈H₄ClF₃O₂
MDL Number:MFCD00052330
:224.57 g/mol
Boiling Pt:94…96 °C/15mmHg
Density:1.435 g/cm³ (25 °C)
Flash Pt:102 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together