
4-(Trifluoromethoxy)benzoic acid hydrazide 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175277-18-6
:
Formula:C₈H₇F₃N₂O₂
MDL Number:MFCD00204234
:220.15 g/mol
Melting Pt:110…120 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together