
4-(Trifluoromethoxy)benzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:330-12-1
:
Formula:C₈H₅F₃O₃
MDL Number:MFCD00002541
:206.12 g/mol
Melting Pt:150…154 °C
EINECS:206-352-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together