
4-(Trifluoromethoxy)benzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:94108-56-2
:
Formula:C₇H₄ClF₃O₃S
MDL Number:MFCD00042408
:260.62 g/mol
Boiling Pt:120…122 °C/15mmHg
Density:1.536 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:302-397-9
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together