
4-(Trifluoromethoxy)benzamide 97%
Catalog # APOSPC7436-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.14 g/mol
Melting Pt:155…156 °C
:GHS07
Storage Temperature:Ambient
CAS Number:456-71-3
Formula:C₈H₆F₃NO₂
MDL Number:MFCD00041384