Order Entry
Czech Republic
ContactUsLinkComponent
2-(Trifluoromethoxy)benzaldehyde 98%
Irritant
2-(Trifluoromethoxy)benzaldehyde 98%
Catalog # APOSPC7434E-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(Trifluoromethoxy)benzaldehyde 98%
Catalog # APOSPC7434E-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :190.12 g/mol
Boiling Pt:77 °C/20mmHg
Density:1.332 g/cm³ (25 °C)
Flash Pt:67.2 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:94651-33-9
Formula:C₈H₅F₃O₂
MDL Number:MFCD00042405