
2-Bromo-5-fluoroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1003-99-2
:
Formula:C₆H₅BrFN
MDL Number:MFCD00070750
Boiling Pt:109…110 °C/15mmHg
Melting Pt:42…47 °C
Flash Pt:110 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:190.01 g/mol
:GHS07
Frequently Bought Together