

5-Fluoro-2-(trifluoromethyl)phenol 98%
Catalog # APOSPC7325-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.1 g/mol
Melting Pt:48…52 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
:GHS07
CAS Number:243459-91-8
Formula:C₇H₄F₄O
MDL Number:MFCD00236302