
4'-Fluoro-3'-methoxyacetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64287-19-0
:
Formula:C₉H₉FO₂
MDL Number:MFCD00272135
Melting Pt:51…53 °C
Flash Pt:100 °C
:168.17 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together