0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
5-(Trifluoromethoxy)oxindole
5-(Trifluoromethoxy)oxindole 98%
5-(Trifluoromethoxy)oxindole 98%
APOSPC7125-1G
:
Apollo Scientific
:
5-(Trifluoromethoxy)oxindole 98%
APOSPC7125-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
217.15 g/mol
:
GHS07
:
Refrigerator
:
403853-48-5
:
C₉H₆F₃NO₂
:
MFCD09027275