Order Entry
Czech Republic
ContactUsLinkComponent
2-Fluoro-1,3-dimethoxy-4-nitrobenzene 98%
Irritant
2-Fluoro-1,3-dimethoxy-4-nitrobenzene 98%
Catalog # APOSPC7106-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Fluoro-1,3-dimethoxy-4-nitrobenzene 98%
Catalog # APOSPC7106-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.1500 g/mol
Melting Pt:58…61 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:155020-44-3
Formula:C₈H₈FNO₄
MDL Number:MFCD09027197