
2-Fluoro-6-(trifluoromethyl)phenylboronic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:313545-34-5
:
Formula:C₇H₅BF₄O₂
MDL Number:MFCD07781239
:207.92 g/mol
Melting Pt:115…119 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together