1-Chloro-1,2-dibromo-1-fluoroethane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:243139-69-7
:
Formula:C₂H₂Br₂ClF
MDL Number:MFCD01320780
Boiling Pt:134…136 °C
:240.3
Storage Temperature:Ambient
Frequently Bought Together