Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
6-Fluoroquinoline

6-Fluoroquinoline

Corrosive
Irritant
Supplier:   Apollo Scientific
6-Fluoroquinoline
Supplier:  Apollo Scientific
Certificates
6-Fluoroquinoline

5 Options Available Below

Product Details & Documents
  • CAS Number:396-30-5
  • Formula:C₉H₆FN
  • MDL Number:MFCD01685512
  • Molecular Weight:147.15 g/mol
  • Boiling Pt:124 °C/30mmHg
  • Density:1.209 g/cm³ (25 °C)
  • Storage Temperature:Refrigerator
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 615,00 Kč
    615,00 Kč
    1 * 1 g
       615,00 Kč

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 697,00 Kč
    697,00 Kč
    1 * 5 g
       697,00 Kč

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 2 220,00 Kč
    2 220,00 Kč
    1 * 25 g
       2 220,00 Kč

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 7 920,00 Kč
    7 920,00 Kč
    1 * 100 g
       7 920,00 Kč

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 38 700,00 Kč
    38 700,00 Kč
    1 * 500 g
       38 700,00 Kč

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.