Order Entry
Czech Republic
ContactUsLinkComponent
3-Fluoro-4-(trifluoromethyl)benzophenone 97%
Irritant
3-Fluoro-4-(trifluoromethyl)benzophenone 97%
Catalog # APOSPC6907-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Fluoro-4-(trifluoromethyl)benzophenone 97%
Catalog # APOSPC6907-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:243128-47-4
Formula:C₁₄H₈F₄O
MDL Number:MFCD00236282