
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:203866-13-1
:
Formula:C₁₀H₁₆FNO₄
MDL Number:MFCD04973957
:233.24 g/mol
Melting Pt:157…162 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together