
Methyl-2,6-dichloro-5-fluoronicotinate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:189281-66-1
:
Formula:C₇H₄Cl₂FNO₂
MDL Number:MFCD02180634
:224.02 g/mol
Boiling Pt:84…86 °C/0.1mmHg
:GHS07
Storage Temperature:Ambient
Frequently Bought Together