
2-(2-Chloro-4-fluorophenyl)-N-hydroxyacetamidine 97%
Catalog # APOSPC6712-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:202.62 g/mol
Melting Pt:149…151 °C
:GHS07
Storage Temperature:Ambient
CAS Number:306937-33-7
Formula:C₈H₈ClFN₂O
MDL Number:MFCD02180957