Order Entry
Czech Republic
ContactUsLinkComponent
5-Fluoro-2-(trifluoromethyl)phenylacetonitrile 98%
Irritant
5-Fluoro-2-(trifluoromethyl)phenylacetonitrile 98%
Catalog # APOSPC6708-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Fluoro-2-(trifluoromethyl)phenylacetonitrile 98%
Catalog # APOSPC6708-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :203.14 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
 :GHS07
CAS Number:242812-09-5
Formula:C₉H₅F₄N
MDL Number:MFCD06247720