
2,3,5,6-Tetrafluoroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:700-17-4
:
Formula:C₆H₃F₄N
MDL Number:MFCD00007644
:165.09 g/mol
Boiling Pt:158 °C
Melting Pt:31…33 °C
Density:1.744 g/cm³ (25 °C)
Flash Pt:62 °C
EINECS:211-840-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together