
Tetrabutylammonium hexafluorophosphate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3109-63-5
:
Formula:C₁₆H₃₆F₆NP
MDL Number:MFCD00011748
:387.43 g/mol
Melting Pt:244…248 °C
EINECS:221-472-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together