Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Iodo-2-(heptafluoropropyl)cyclohexane (E/Z)
1-Iodo-2-(heptafluoropropyl)cyclohexane (E/Z)
1-Iodo-2-(heptafluoropropyl)cyclohexane (E/Z)
1-Iodo-2-(heptafluoropropyl)cyclohexane (E/Z)
APOSPC6544-250MG
:
Apollo Scientific
:
1-Iodo-2-(heptafluoropropyl)cyclohexane (E/Z)
APOSPC6544-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
378.07 g/mol
:
48…50 °C/1.3mmHg
:
1.8 g/cm³ (25 °C)
:
Ambient
:
2728-75-8
:
C₉H₁₀F₇I
:
MFCD02183508