Order Entry
Czech Republic
ContactUsLinkComponent
Sodium hexafluorophosphate, anhydrous 98%
Corrosive
Irritant
Sodium hexafluorophosphate, anhydrous 98%
Catalog # APOSPC6510-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
Sodium hexafluorophosphate, anhydrous 98%
Catalog # APOSPC6510-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Melting Pt:>200 °C
Density:2.369 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3260
ADR:8,II
 :167.95 g/mol
EINECS:244-333-1
 :GHS05
 :GHS07
CAS Number:21324-39-0
Formula:F₆NaP
MDL Number:MFCD00011122
Melting Pt:>200 °C