
5-Chloro-4-(4-fluorophenyl)-1,2,3-thiadiazole ≥95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:77414-44-9
:
Formula:C₈H₄ClFN₂S
MDL Number:MFCD00173564
Melting Pt:109…111 °C
:214.65 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together