
2-(2-Hydroxyethyl)benzotrifluoride
Catalog # APOSPC6258-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Boiling Pt:184…186 °C
Melting Pt:–3 to –2 °C
Density:1.197 g/cm³ (25 °C)
Flash Pt:103 °C
:190.17 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:94022-96-5
Formula:C₉H₉F₃O
MDL Number:MFCD00003540
Melting Pt:–3…–2 °C