
4-Bromo-3-chlorobenzotrifluoride 98%
Catalog # APOSPC6198-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:259.45 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:402-04-0
Formula:C₇H₃BrClF₃
MDL Number:MFCD00673985