Order Entry
Czech Republic
ContactUsLinkComponent
8H-Perfluorooctyl iodide
Irritant
8H-Perfluorooctyl iodide
Katalogové číslo APOSPC6171B-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
8H-Perfluorooctyl iodide
Katalogové číslo APOSPC6171B-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :527.98 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:79162-64-4
Formula:C₈HF₁₆I
MDL Number:MFCD00236712