2-Chloro-3,6-difluoro-4-hydroxybenzenecarboxylic acid
Catalog # APOSPC6148-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:208.55 g/mol
Storage Temperature:Ambient
CAS Number:749230-42-0
Formula:C₇H₃ClF₂O₃
MDL Number:MFCD12025951