
2-Methyl-4-(trifluoromethyl)-1,3-thiazole-5-carbonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:117724-64-8
:
Formula:C₆H₃ClF₃NOS
MDL Number:MFCD11845754
:229.61 g/mol
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together