Order Entry
Czech Republic
ContactUsLinkComponent
4-Nitro-3-(trifluoromethyl)anisole 98%
Irritant
4-Nitro-3-(trifluoromethyl)anisole 98%
Catalog # APOSPC6093-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Nitro-3-(trifluoromethyl)anisole 98%
Catalog # APOSPC6093-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :221.1334 g/mol
Melting Pt:48 °C
Flash Pt:>110 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:344-39-8
Formula:C₈H₆F₃NO₃
MDL Number:MFCD00053611