
1H,1H,2H-Perfluoro-1-hexene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19430-93-4
:
Formula:C₆H₃F₉
MDL Number:MFCD00042338
:246.08 g/mol
Boiling Pt:59 °C
Density:1.457 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:243-053-7
UN:1993
ADR:3,II
:GHS02
Frequently Bought Together