7-Hydroxy-4-(3-trifluoromethylphenyl)coumarin 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:386704-09-2
:
Formula:C₁₆H₉F₃O₃
MDL Number:MFCD01941326
:306.24 g/mol
Melting Pt:>200 °C (dec.)
Storage Temperature:Ambient
Frequently Bought Together