
Methyl-4-bromo-3-(trifluoromethyl)benzoate 98%
Catalog # APOSPC5880-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:283.0420 g/mol
Melting Pt:55…57 °C
:GHS07
Storage Temperature:Ambient
CAS Number:107317-58-8
Formula:C₉H₆BrF₃O₂
MDL Number:MFCD10566409