Order Entry
Czech Republic
ContactUsLinkComponent
4-(Trifluoromethyl-1,3-thiazole-2-carboxylic acid 98%
Irritant
4-(Trifluoromethyl-1,3-thiazole-2-carboxylic acid 98%
Catalog # APOSPC5807-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(Trifluoromethyl-1,3-thiazole-2-carboxylic acid 98%
Catalog # APOSPC5807-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :197.14 g/mol
Melting Pt:109…111 °C
Flash Pt:>110 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:944900-55-4
Formula:C₅H₂F₃NO₂S
MDL Number:MFCD07377490