
Pentafluoropropionic anhydride 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:356-42-3
:
Formula:C₆F₁₀O₃
MDL Number:MFCD00000429
:310.05 g/mol
Boiling Pt:73…74 °C
Density:1.571 g/cm³ (25 °C)
Storage Temperature:Refrigerator
EINECS:206-604-2
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together