Order Entry
Czech Republic
ContactUsLinkComponent
1H,2H-Octafluorocyclopentane
Irritant
1H,2H-Octafluorocyclopentane
Catalog # APOSPC5746-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1H,2H-Octafluorocyclopentane
Catalog # APOSPC5746-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.06 g/mol
Boiling Pt:79 °C
Density:1.68 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:828-35-3
Formula:C₅H₂F₈
MDL Number:MFCD03094303