
2-(Pentafluorophenyl)-2-propanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:715-31-1
:
Formula:C₉H₇F₅O
MDL Number:MFCD00039613
:226.15 g/mol
Boiling Pt:67…68 °C/5mmHg
Melting Pt:78…80 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together