Order Entry
Czech Republic
ContactUsLinkComponent
Tetramethylammonium fluoride tetrahydrate 97%
Irritant
Tetramethylammonium fluoride tetrahydrate 97%
Catalog # APOSPC5687-500G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Tetramethylammonium fluoride tetrahydrate 97%
Catalog # APOSPC5687-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :165.21 g/mol
Melting Pt:39…42 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:17787-40-5
Formula:C₄H₁₂FN·4H₂O
MDL Number:MFCD00149968