
3,4-Difluorobenzylamine 94%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:72235-53-1
:
Formula:C₇H₇F₂N
MDL Number:MFCD00010145
:143.14 g/mol
Density:1.21 g/cm³ (25 °C)
Flash Pt:79 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together