
3-((Trifluoromethyl)thio)benzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:946-65-6
:
Formula:C₈H₅F₃O₂S
MDL Number:MFCD00236337
:222.19 g/mol
Melting Pt:71…74 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together