

4-(Trifluoromethyl)-N,N-dimethylbenzamide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:25771-21-5
:
Formula:C₁₀H₁₀F₃NO
MDL Number:MFCD16618942
Melting Pt:95…96 °C
:217.1877 g/mol
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together