Order Entry
Czech Republic
ContactUsLinkComponent
Pentafluorobenzyl bromide 98%
Corrosive
Irritant
Pentafluorobenzyl bromide 98%
Catalog # APOSPC5550-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
Pentafluorobenzyl bromide 98%
Catalog # APOSPC5550-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :260.99 g/mol
Boiling Pt:174…175 °C
Melting Pt:19…20 °C
Density:1.728 g/cm³ (25 °C)
Flash Pt:83 °C
Storage Temperature:Ambient
EINECS:217-182-4
UN:3265
ADR:8,II
 :GHS05
 :GHS07
CAS Number:1765-40-8
Formula:(C₆F₅)CH₂Br
MDL Number:MFCD00000299