Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromotetrafluoropropionic acid 97%
Corrosive
Irritant
2-Bromotetrafluoropropionic acid 97%
Catalog # APOSPC5515-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2-Bromotetrafluoropropionic acid 97%
Catalog # APOSPC5515-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :224.94 g/mol
Boiling Pt:60 °C/22mmHg
Storage Temperature:Ambient
UN:3265
ADR:8,III
 :GHS05
 :GHS07
CAS Number:13859-31-9
Formula:C₃HBrF₄O₂
MDL Number:MFCD02183518