
2-Hydroxy-6-(trifluoromethyl)nicotinamide
Catalog # APOSPC5476-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.13 g/mol
Melting Pt:220…223 °C
:GHS07
Storage Temperature:Ambient
CAS Number:116548-03-9
Formula:C₇H₅F₃N₂O₂
MDL Number:MFCD00243729