
6-(Trifluoromethyl)nicotinoyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:358780-13-9
:
Formula:C₇H₃ClF₃NO
MDL Number:MFCD01862645
:209.5530 g/mol
Boiling Pt:54 °C/2mmHg
Density:1.438 g/cm³ (25 °C)
Flash Pt:108.9 °C
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together