
2-Chloro-6-(trifluoromethyl)nicotinonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:386704-06-9
:
Formula:C₇H₂ClF₃N₂
MDL Number:MFCD01862660
:206.5500 g/mol
Boiling Pt:74…76 °C/0.8mmHg
Melting Pt:36…38 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together